missing translation for 'onlineSavingsMsg'
Learn More

L-Methionine-13C, TRC

Product Code. 30481325
Change view
Click to view available options
Quantity:
0.25 mg
0.5 mg
2.5 mg
Unit Size:
0.25mg
0.5mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481325 0.5 mg 0.5mg
30467913 0.25 mg 0.25mg
30469503 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481325 Supplier Toronto Research Chemicals Supplier No. TRCM2605650.5MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49705-26-2
Molecular Formula C413CH11NO2S
Molecular Weight (g/mol) 150.2
Synonym L-Methionine-methyl-13C (9CI, ACI), L-(S-Methyl-13C)methionine, L-Methionine-methyl-13C, [Me-13C]-L-methionine, L-Methionine-13C, L-Methionine-13C (S-methyl-13C)
SMILES [13CH3]SCC[C@H](N)C(O)=O

Specifications

Quantity 0.5 mg
InChI Formula InChI=1S/C5H11NO2S/c1-9-3-2-4(6)5(7)8/h4H,2-3,6H2,1H3,(H,7,8)/t4-/m0/s1/i1+1
Chemical Name or Material L-Methionine-13C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.