missing translation for 'onlineSavingsMsg'
Learn More

L-Menthone, 97%

Product Code. 11498840
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11498840

Brand: Thermo Scientific Alfa Aesar A13679.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14073-97-3 | C10H18O | 154.25 g/mol

L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Menthone is used in perfumery and cosmetics for its characteristic aromatic and minty odor. And it is also used in synthesis of agrochemicals, and dyestuff.

Solubility
Soluble in ether, alcohol, benzene, acetone.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14073-97-3
Molecular Formula C10H18O
Molecular Weight (g/mol) 154.25
MDL Number MFCD00001634,MFCD00136033
InChI Key NFLGAXVYCFJBMK-BDAKNGLRSA-N
Synonym --menthone, l-menthone, menthone, trans-menthone, p-menthone, neomenthone, menthone racemic, trans-p-menthan-3-one, 2s,5r-2-isopropyl-5-methylcyclohexanone, l-menthan-3-one
PubChem CID 26447
ChEBI CHEBI:15410
IUPAC Name (2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-one
SMILES CC(C)[C@@H]1CC[C@@H](C)CC1=O

Specifications

Density 0.893
Boiling Point 208°C to 210°C
Flash Point 72°C (162°F)
Assay Percent Range 97%
Refractive Index 1.45
Quantity 100 g
Beilstein 3648743
Merck Index 14,5838
Solubility Information Soluble in ether,alcohol,benzene,acetone.
Optical Rotation −25° (Neat)
Formula Weight 154.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material L-Menthone
Show More Show Less
compliance-icons
Product Identifier
  • L-Menthone
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1B
Hazard Statement
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contains : L-Menthone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.