missing translation for 'onlineSavingsMsg'
Learn More

L-Menthol, 99%

Product Code. 11418713
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11418713 250 g 250g
11408713 50 g 50g
11498703 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11418713 Supplier Thermo Scientific Alfa Aesar Supplier No. A10474.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2216-51-5 | C10H20O | 156.269 g/mol

L-Menthol is used as a cooling agent that strongly activates TRPM8.(Transient Receptor Potential Cation Channel, Subfamily M, Member 8 is a Protein Coding gene). It is used as analgesic (topical), antipruritic agent. It is used as: refreshing agent, food flavor, cool and antipruritic drug, carminative drug. Menthol crystals is used for pers onal care and cosmetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in 100% ethanol, methanol (100 mg/ml), ether, and chloroform. Insoluble in water.

Notes
Store at room temperature. Incompatible with Strong oxidizing agents, phenols, potassium permanganate, chromium trioxide, chloral hydrate, thymol, pyrogallol, resorcinol, camphor, butylchloral hydrate, betanaphthol, exalgine.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2216-51-5
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00062979
InChI Key NOOLISFMXDJSKH-KXUCPTDWSA-N
Synonym l-menthol, --menthol, levomenthol, menthomenthol, l---menthol, hexahydrothymol, menthacamphor, peppermint camphor, 1r,2s,5r-2-isopropyl-5-methylcyclohexanol, u.s.p. menthol
PubChem CID 16666
ChEBI CHEBI:15409
IUPAC Name (1R,2S,5R)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Melting Point 42°C to 45°C
Density 0.89
Boiling Point 212°C to 216°C
Flash Point 93°C (199°F)
Quantity 250 g
Beilstein 1902293
Merck Index 14,5837
Solubility Information Soluble in 100% ethanol,methanol (100mg/ml),ether,and chloroform. Insoluble in water.
Optical Rotation −50° (c=10 in ethanol)
Formula Weight 156.27
Percent Purity 99%
Chemical Name or Material L-Menthol
Show More Show Less
compliance-icons
Product Identifier
  • L-Menthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P314-Get medical advice/attention if you feel unwell.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: L-Menthol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.