missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Malic acid, 97%, Thermo Scientific Chemicals

Catalog Number 11464694
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy
97-67-6
C4H6O5
134.087
MFCD00064213
BJEPYKJPYRNKOW-REOHCLBHSA-N
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
222656
CHEBI:30797
(2S)-2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O
This item is not returnable. View return policy
97-67-6
C4H6O5
134.087
MFCD00064213
BJEPYKJPYRNKOW-REOHCLBHSA-N
l-malic acid, l---malic acid, s-2-hydroxysuccinic acid, 2s-2-hydroxybutanedioic acid, l--malic acid, apple acid, --malic acid, l-apple acid, s-malic acid, s-2-hydroxybutanedioic acid
222656
CHEBI:30797
(2S)-2-hydroxybutanedioic acid
C(C(C(=O)O)O)C(=O)O

L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-(-)-Malic acid is used as Selective α-amino protecting reagent for amino acid derivatives. Versatile synthon for the preparation of chiral compounds including κ-opioid rece. It also acts as active ingredient in many sour or tart foods. It is used as synthesizing disincrustant and fluorescent whitening agent. It aids in the production of polyester and alcohol acid resins.

Solubility
Soluble in acetone, dioxane, water, methanol and ethanol. Insoluble in benzene

Notes
Incompatible with Bases, Oxidizing agents, Reducing agents, Alkali metals .
TRUSTED_SUSTAINABILITY
Melting Point 100°C to 106°C
Assay Percent Range 97%
Beilstein 1723541
Merck Index 14,5707
Solubility Information Soluble in acetone, dioxane, water, methanol and ethanol; Insoluble in benzene
Formula Weight 134.09
Percent Purity 97%
Density 1.6
Quantity 100 g
Flash Point 220°C (428°F)
Chemical Name or Material L-(-)-Malic Acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'