missing translation for 'onlineSavingsMsg'
Learn More

L-m-Tyrosine, TRC

Product Code. 30451657
Change view
Click to view available options
Quantity:
50 mg
250 mg
5 g
Unit Size:
250mg
50mg
5g
Product Code. Quantity unitSize
30451657 50 mg 50mg
30451655 5 g 5g
30453534 250 mg 250mg
3 options
This item is not returnable. View return policy

Product Code. 30451657

Brand: Toronto Research Chemicals TRCT91001050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 587-33-7
Formula Weight 181.0739
InChI Formula InChI=1S/C9H11NO3/c10-8(9(12)13)5-6-2-1-3-7(11)4-6/h1-4,8,11H,5,10H2,(H,12,13)/t8-/m0/s1
IUPAC Name (2S)-2-amino-3-(3-hydroxyphenyl)propanoic acid
Molecular Formula C9 H11 N O3
Molecular Weight (g/mol) 181.19
SMILES N[C@@H](Cc1cccc(O)c1)C(=O)O
Synonym L-Phenylalanine, 3-hydroxy-, m-Tyrosine, L- (8CI), 3-Hydroxy-L-phenylalanine, (2S)-2-Amino-3-(3-hydroxyphenyl)propanoic acid, (S)-2-Amino-3-(3-hydroxyphenyl)propanoic acid, 3-(m-Hydroxyphenyl)alanine, 3-Hydroxyphenylalanine, L-3-Hydroxyphenylalanine, L-m-Tyrosine, L-meta-Tyrosine, m-L-Tyrosine, (S)-α-Amino-β-(m-hydroxyphenyl)propionic acid
Chemical Name or Material L-m-Tyrosine

Specifications

Quantity 50 mg
Shipping Condition Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.