missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Monohydrate (>90%), TRC

Product Code. 30427374
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30427374 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30427374 Supplier Toronto Research Chemicals Supplier No. TRCS2588631G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lysine Monohydrate
CAS 39665-12-8
Purity Grade Notes HPLC
Molecular Formula C6 H14 N2 O2 . H2 O
Quantity 1 g
Synonym L-Lysine Monohydrate, L-Lysine','hydrate (1:1), L-Lysine','monohydrate (9CI)
InChI Formula InChI=1S/C6H14N2O2.H2O/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H2/t5-;/m0./s1
SMILES O.NCCCC[C@H](N)C(=O)O
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrate
Molecular Weight (g/mol) 164.203
Formula Weight 164.116
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.