missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Hydrochloride (White Powder), Fisher BioReagents

Product Code. 10438733
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10438733 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10438733 Supplier Fisher BioReagents Supplier No. BP386100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 263°C
Ammonium 0.02% max.
CAS 657-27-2
Arsenic (As) 1ppm max.
Color White
Chloride 19.12 to 19.51%
Heavy Metals (as Pb) 10ppm max.
Identification Pass Test
Assay Percent Range 98.5 to 100.5 %
Loss on Drying 0.4% max.
Other Amino Acids Chromatographically not detectable
Molecular Formula C6H15ClN2O2
Packaging Glass Bottle
MDL Number MFCD00064564 MFCD00081870
Residue after Ignition 0.1% max.
Specific Rotation 20.7 to 21.5° (+ or -)
Merck Index 15, 5697
State of Solution (%T) 98.0% min.
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
Alpha Vector Lysine Hydrochloride
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
SMILES C(CCN)CC(C(=O)O)N.Cl
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
Molecular Weight (g/mol) 182.648
Quantity 100 g
PubChem CID 69568
ChEBI CHEBI:53633
Formula Weight 182.65
Percent Purity 98.5 to 100.5%
Physical Form Solid
Chemical Name or Material L-Lysine Hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.