missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Hydrochloride, TRC

Product Code. 30134982
Change view
Click to view available options
Quantity:
100 g
250 g
500 g
Unit Size:
100g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134982 500 g 500g
30007180 100 g 100g
30012400 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134982 Supplier Toronto Research Chemicals Supplier No. TRCL468915500G
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6 H14 N2 O2 . Cl H
Molecular Weight (g/mol) 182.65
Synonym L-Lysine, hydrochloride (1:1), L-Lysine, monohydrochloride (9CI), Lysine, monohydrochloride, L- (8CI), Bovi-Lysine, Darvyl, L-(+)-Lysine monohydrochloride, L-Gen, L-Lysine hydrochloride, L-Lysine monohydrochloride, Lyamine, Lysine hydrochloride, Lysine monohydrochloride, Lysion, NSC 9253, Relys, Lysine hydrochloride, (S)-2,6-diaminohexanoic acid hydrochloride
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES Cl.NCCCC[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C6H14N2O2.ClH/c7-4-2-1-3-5(8)6(9)10;/h5H,1-4,7-8H2,(H,9,10);1H/t5-;/m0./s1
Quantity 500 g
Formula Weight 182.0822
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Lysine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.