missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Hydrochloride, 99+%, MP Biomedicals

Product Code. 11491901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11491901 500 g 500g
11481901 100 g 100g
11401911 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11491901 Supplier MP Biomedicals Supplier No. 0219469790
 Restricted Item
Estimated Shipment: 12-02-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 657-27-2 | C6H15ClN2O2 | 182.65 g/mol

Chemical Identifiers

CAS 657-27-2
Molecular Formula C6H15ClN2O2
Molecular Weight (g/mol) 182.648
InChI Key BVHLGVCQOALMSV-JEDNCBNOSA-N
Synonym l-lysine hydrochloride, l-lysine monohydrochloride, lysine hydrochloride, h-lys-oh.hcl, l-lysine, monohydrochloride, lyamine, darvyl, lysion, s-2,6-diaminohexanoic acid hydrochloride, enisyl
PubChem CID 69568
ChEBI CHEBI:53633
IUPAC Name (2S)-2,6-diaminohexanoic acid;hydrochloride
SMILES C(CCN)CC(C(=O)O)N.Cl

Specifications

Quantity 500 g
Formula Weight 182.6
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Lysine, 99+%, Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.