missing translation for 'onlineSavingsMsg'
Learn More

L-Lysine Dihydrochloride 99%, Thermo Scientific Chemicals

Catalog Number 10197080
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
657-26-1
C6H16Cl2N2O2
219.11
MFCD00066389
JBBURJFZIMRPCZ-XRIGFGBMSA-N
l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
69567
Cl.Cl.NCCCC[C@H](N)C(O)=O
This item is not returnable. View return policy
657-26-1
C6H16Cl2N2O2
219.11
MFCD00066389
JBBURJFZIMRPCZ-XRIGFGBMSA-N
l-lysine dihydrochloride, h-lys-oh.2hcl, l-lysine, dihydrochloride, s-2,6-diaminohexanoic acid dihydrochloride, unii-90yfr6u9qc, l-+-lysine dihydrochloride, 90yfr6u9qc, 2s-2,6-diaminohexanoic acid dihydrochloride, lysine, chloride, chloride, l-lysine, hydrochloride 1:2
69567
Cl.Cl.NCCCC[C@H](N)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 202.0°C
Color White to Yellow
Assay Percent Range 99%
Infrared Spectrum Authentic
Linear Formula NH2(CH2)4CH(NH2)COOH·2HCl
Loss on Drying 0.5% max. (105°C, 3 hrs)
Merck Index 14, 5636
Packaging Glass Bottle
IUPAC Name (2S)-2,6-diaminohexanoic acid;dihydrochloride
Specific Rotation + 14.00
Formula Weight 219.11
Percent Purity 99%
Specific Rotation Condition +14.00 (18.00°C c=2,water)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material L-Lysine dihydrochloride, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'