missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, TRC

Product Code. 30132838
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30132838 25 g 25g
17978324 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132838 Supplier Toronto Research Chemicals Supplier No. TRCL33011025G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6 H13 N O2
Molecular Weight (g/mol) 131.17
Synonym L-Leucine, Leucine, L- (8CI), (2S)-2-Amino-4-methylpentanoic acid, (S)-(+)-Leucine, (S)-2-Amino-4-methylpentanoic acid, (S)-2-Amino-4-methylvaleric acid, (S)-Leucine, L-(+)-Leucine, L-2-Amino-4-methylpentanoic acid, L-Norvaline, 4-methyl-, L-α-Aminoisocaproic acid, Leu, Leucine, Pentanoic acid, 2-amino-4-methyl-, (S)-, NSC 46709
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C6H13NO2/c1-4(2)3-5(7)6(8)9/h4-5H,3,7H2,1-2H3,(H,8,9)/t5-/m0/s1
Quantity 25 g
Formula Weight 131.0946
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Leucine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.