missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, MP Biomedicals

Product Code. 11401901 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
Product Code. Quantity unitSize
11401901 25 g 25g
11411901 100 g 100g
11421901 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11401901

Brand: MP Biomedicals 0219469425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 61-90-5
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00002617
InChI Key ROHFNLRQFUQHCH-MLHKIVSYNA-N
Synonym l-leucine, leucine, s-leucine, s-2-amino-4-methylpentanoic acid, h-leu-oh, 2s-2-amino-4-methylpentanoic acid, s-+-leucine, leucin, l-norvaline, 4-methyl, s-2-amino-4-methylvaleric acid
PubChem CID 6106
ChEBI CHEBI:15603
IUPAC Name (2S)-2-amino-4-methylpentanoic acid
SMILES CC(C)C[C@H](N)C(O)=O

Specifications

Quantity 25 g
Formula Weight 131.2
Grade Cell Culture
Physical Form Crystals
Chemical Name or Material L-Leucine, Substantially free of isoleucine and methionine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.