missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Leucine methyl ester hydrochloride, 98%

Product Code. 10571751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571751 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10571751 Supplier Thermo Scientific Chemicals Supplier No. 125130250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7517-19-3 | C7H16ClNO2 | 181.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7517-19-3
Melting Point 145.0°C to 155.0°C
Color White
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H16ClNO2
Linear Formula (CH3)2CHCH2CH(NH2)CO2CH3·HCl
MDL Number MFCD00012494
Quantity 25 g
Beilstein 04, I, 518
Specific Rotation Condition + 20.00 (20.00°C c=4.5,MeOH)
Specific Rotation + 20.00
Synonym l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride
Solubility Information Solubility in water: soluble in water. Other solubilities: 5% soluble in methanol
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
SMILES Cl.COC(=O)[C@@H](N)CC(C)C
IUPAC Name methyl (2S)-2-amino-4-methylpentanoate;hydrochloride
Molecular Weight (g/mol) 181.66
PubChem CID 2723695
Formula Weight 181.66
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Leucine methyl ester hydrochloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.