missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Product Code. 11492697
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11492697 25 g 25g
11402707 100 g 100g
11412707 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492697 Supplier Thermo Scientific Alfa Aesar Supplier No. A14897.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7517-19-3
Molecular Formula C7H16ClNO2
Molecular Weight (g/mol) 181.66
MDL Number MFCD00012494
InChI Key DODCBMODXGJOKD-HQFIOVGONA-N
Synonym l-leucine methyl ester hydrochloride, h-leu-ome.hcl, methyl l-leucinate hydrochloride, s-methyl 2-amino-4-methylpentanoate hydrochloride, h-leu-ome. hcl, leucine methyl ester hydrochloride, h-leu-ome hydrochloride, l-leucine methyl ester hydrochloride;h-leu-ome.hcl, s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride, l-leucine, methyl ester, monohydrochloride
PubChem CID 2723695
IUPAC Name methyl (2S)-2-amino-4-methylpentanoate;hydrochloride
SMILES Cl.COC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 149°C to 151°C
Beilstein 3595133
Quantity 25 g
Formula Weight 181.66
Optical Rotation +13° (c=2 in Water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Leucine methyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.