missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine, MedChemExpress

Product Code. 16419096
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16419096 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16419096 Supplier Medchem Express Supplier No. HYN0486
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Leucine is an essential branched-chain amino acid (BCAA), which activates the mTOR signaling pathway.

L-Leucine is an essential branched-chain amino acid (BCAA), which activates the mTOR signaling pathway.

Specifications

Chemical Name or Material L-Leucine
CAS 61-90-5
Color White
Purity Grade Notes Research
Molecular Formula C6H13NO2
Solubility Information H2O : 8.33 mg/mL (63.51 mM; Need ultrasonic)
SMILES N[C@@H](CC(C)C)C(O)=O
Molecular Weight (g/mol) 131.17
Quantity 100 mg
Formula Weight 131.17
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.