missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine ethyl ester hydrochloride, 99%, Thermo Scientific™

Product Code. 11945391 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11945391 5 g Glass bottle 5g
11965391 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11945391 Supplier Thermo Scientific Acros Supplier No. 442710050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2743-40-0
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00034879
InChI Key QIGLJVBIRIXQRN-ZETCQYMHSA-N
Synonym l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl
PubChem CID 11658447
SMILES CCOC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 135°C to 137°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Specific Rotation 15
IUPAC Name ethyl (2S)-2-amino-4-methylpentanoate
Formula Weight 195.69
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material L-Leucine ethyl ester hydrochloride
Show More Show Less
Product Identifier
  • L-Leucine ethyl ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.