missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine ethyl ester hydrochloride, 97%

Product Code. 11337936
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337936 25 g 25g
11327936 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337936 Supplier Thermo Scientific Alfa Aesar Supplier No. L13147.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2743-40-0 | C8H17NO2 | 159.23 g/mol

L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Leucine ethyl ester hydrochloride is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2743-40-0
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00034879
InChI Key QIGLJVBIRIXQRN-ZETCQYMHSA-N
Synonym l-leucine ethyl ester hydrochloride, ethyl l-leucinate hydrochloride, h-leu-oet.hcl, s-ethyl 2-amino-4-methylpentanoate hydrochloride, l-leucine ethyl ester hcl, h-leu-oet inverted exclamation mark currencyhcl, leucine, ethyl ester hydrochloride, h-leu-oet, leucine ethyl ester hydrochloride, leu-oet hcl
PubChem CID 11658447
SMILES CCOC(=O)[C@@H](N)CC(C)C

Specifications

Melting Point 134°C to 137°C
Beilstein 3994312
Solubility Information Soluble in water.
IUPAC Name ethyl (2S)-2-amino-4-methylpentanoate;hydrochloride
Quantity 25 g
Odor Odorless
Formula Weight 195.69
Optical Rotation +18° (c=5 in Ethanol)
Percent Purity 97%
Chemical Name or Material L-Leucine ethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.