missing translation for 'onlineSavingsMsg'
Learn More

L-Leucine-4-nitroanilide, 99%, Thermo Scientific Chemicals

Product Code. 11464034
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464034 1 g 1g
11474034 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464034 Supplier Thermo Scientific Alfa Aesar Supplier No. A11994.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used to detect/evaluate functionality of amino peptidases. The compound is used in an ELISA assay to detect biological functionality of chicken amino peptidase produced by E. coli. It is also employed to characterize the native IRAP (placental leucine amino peptidase). Leu-pNA, Chromogenic Substrate is documented for use with pure samples as well as cell extracts in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to detect/evaluate functionality of amino peptidases. The compound is used in an ELISA assay to detect biological functionality of chicken amino peptidase produced by E. coli. It is also employed to characterize the native IRAP (placental leucine amino peptidase). Leu-pNA, Chromogenic Substrate is documented for use with pure samples as well as cell extracts in vitro.

Solubility
Soluble in DMSO (>100 mM), and methanol (>25 mM).

Notes
Store away from oxidizing agents. Keep the container tightly closed in a well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4178-93-2
Molecular Formula C12H17N3O3
Molecular Weight (g/mol) 251.29
MDL Number MFCD00063681
InChI Key AXZJHDNQDSVIDR-NSHDSACASA-N
Synonym l-leucine-4-nitroanilide, h-leu-pna, l-leucine-p-nitroanilide, s-2-amino-4-methyl-n-4-nitrophenyl pentanamide, 2s-2-amino-4-methyl-n-4-nitrophenyl pentanamide, pentanamide, 2-amino-4-methyl-n-4-nitrophenyl-, s, l-leucyl-p-nitroanilide, pentanamide, 2-amino-4-methyl-n-4-nitrophenyl-, 2s, h-leu-pna.hcl, l-leu p-nitroanilide
PubChem CID 2733285
IUPAC Name (2S)-2-amino-4-methyl-N-(4-nitrophenyl)pentanamide
SMILES CC(C)C[C@H](N)C(=O)NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 89°C to 90°C
Beilstein 2218433
Solubility Information Soluble in DMSO (>100 mM),and methanol (>25 mM).
Quantity 1 g
Formula Weight 251.29
Percent Purity 99%
Chemical Name or Material L-Leucine-4-nitroanilide
compliance-icons
Product Identifier
  • L-Leucine-4-nitroanilide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: L-Leucine-4-nitroanilide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.