missing translation for 'onlineSavingsMsg'
Learn More

L-Leucinamide hydrochloride, 99%

Product Code. 10006203 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10006203 1 g Glass bottle 1g
10267030 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10006203 Supplier Thermo Scientific Acros Supplier No. 302980010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10466-61-2 | C6H15ClN2O | 166.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10466-61-2
Molecular Formula C6H15ClN2O
Molecular Weight (g/mol) 166.65
MDL Number MFCD00013012
InChI Key VSPSRRBIXFUMOU-XZNNNFJINA-N
Synonym l-leucinamide hydrochloride, h-leu-nh2.hcl, s-2-amino-4-methylpentanamide hydrochloride, l-leucinamide hcl, 2s-2-amino-4-methylpentanamide hydrochloride, pentanamide, 2-amino-4-methyl-, monohydrochloride, 2s, c6h14n2o.hcl, l-leucine amide hydrochloride, leu-nh2.hcl, h-d-leu-nh hcl
PubChem CID 2733199
IUPAC Name (2S)-2-amino-4-methylpentanamide;hydrochloride
SMILES Cl.CC(C)C[C@H](N)C(N)=O

Specifications

Melting Point 244°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2CH(NH2)CONH2·HCl
Quantity 1 g
Specific Rotation Condition +9° (20°C c=1,H2O)
Specific Rotation + 9
Formula Weight 166.65
Percent Purity 99%
Physical Form Powder
Chemical Name or Material L-Leucinamide hydrochloride, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.