missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lactic acid lithium salt, Thermo Scientific™

Product Code. 15845308 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15845308 1 kg Plastic bottle 1kg
15855168 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15845308 Supplier Thermo Scientific Supplier No. J18160.A1
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27848-80-2
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
MDL Number MFCD00065399
InChI Key GKQWYZBANWAFMQ-DKWTVANSSA-M
Synonym lithium s-2-hydroxypropanoate, l-lactic acid lithium salt, lithium l-lactate, s-2-hydroxypropionic acid lithium salt, sarcolactic acid lithium salt, lithium 1+ l-+-lactate, lithiuml-lactate, lithotab l-lactate, lactic acid lithium salt, c3h5o3.li
PubChem CID 23687877
IUPAC Name lithium;(2S)-2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Melting Point 253-255°
Packaging Plastic bottle
Quantity 1 kg
Beilstein 3568332
Formula Weight 96.01
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Lactic acid lithium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.