missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Lactic acid lithium salt, 99%, MP Biomedicals™

Product Code. 11388989 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388989 10 g 10g
11398989 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388989 Supplier MP Biomedicals Supplier No. 0210213410
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 867-55-0
Molecular Formula C3H5LiO3
Molecular Weight (g/mol) 96.01
InChI Key GKQWYZBANWAFMQ-UHFFFAOYSA-M
Synonym lithium lactate, lithium 2-hydroxypropanoate, lactic acid, lithium salt, dl-lactic acid lithium salt, propanoic acid, 2-hydroxy-, monolithium salt, lithium 1+ ion lactate, l-+-lactic acid lithium salt, lithotab lactate, acmc-20akmj, lithium s-lactate
PubChem CID 23673452
IUPAC Name lithium;2-hydroxypropanoate
SMILES [Li+].CC(C(=O)[O-])O

Specifications

Color White
Quantity 10 g
Physical Form Powder
Formula Weight 96.1
Percent Purity 99%
Chemical Name or Material L-(+)-Lactic acid lithium salt
compliance-icons
Product Identifier
  • L-(+)-Lactic Acid, free acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.