missing translation for 'onlineSavingsMsg'
Learn More

L-Lactic Acid, 90%, TRC

Product Code. 30138060
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138060 10 g 10g
30135744 5 g 5g
17991155 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138060 Supplier Toronto Research Chemicals Supplier No. TRCL11350010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-33-4
Molecular Formula C3 H6 O3
Molecular Weight (g/mol) 90.08
Synonym S-Lactic acid, (S)-2-hydroxypropanoic acid
IUPAC Name (2S)-2-hydroxypropanoic acid
SMILES C[C@H](O)C(=O)O

Specifications

InChI Formula InChI=1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)/t2-/m0/s1
Formula Weight 90.0317
Quantity 10 g
Chemical Name or Material L-(+)-Lactic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.