missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L(+)-Lactic acid, 90% solution in water
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
missing translation for 'unitSize'
1 L
250mL
5 L
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 125.0°C |
| Color | Colorless to Yellow |
| Linear Formula | CH3CH(OH)CO2H |
| Beilstein | 03, IV, 633 |
| Fieser | 15,181 |
| Merck Index | 15, 5385 |
| Solubility Information | Solubility in water: soluble. Other solubilities: soluble in alcohol, furfurol and ether, practically insoluble in chloroform and carbon, disulfide |
| Formula Weight | 90.08 |
| Percent Purity | ≥88% |
| Density | 1.2000g/mL |
| Show More |
Safety and Handling
Product Identifier
- L(+)-Lactic acid, 90% solution in water
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction