missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L,L-Dityrosine Dihydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | L,L-Dityrosine Dihydrochloride |
| CAS | 221308-01-6 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18 H20 N2 O6 . 2 Cl H |
| Quantity | 2.5 mg |
| Synonym | (α3S,α3'S)-α3,α3'-Diamino-6,6'-dihydroxy-[1,1'-biphenyl]-3,3'-dipropanoic Acid Dihydrochloride, [S-(R*,R*)]- α,α'-Diamino-6,6'-dihydroxy-[1,1'-biphenyl]-3,3'- |
| InChI Formula | InChI=1S/C18H20N2O6.2ClH/c19-13(17(23)24)7-9-1-3-15(21)11(5-9)12-6-10(2-4-16(12)22)8-14(20)18(25)26;;/h1-6,13-14,21-22H,7-8,19-20H2,(H,23,24)(H,25,26);2*1H/t13-,14-;;/m0../s1 |
| SMILES | Cl.Cl.N[C@@H](Cc1ccc(O)c(c1)c2cc(C[C@H](N)C(=O)O)ccc2O)C(=O)O |
| IUPAC Name | (2S)-2-amino-3-[3-[5-[(2S)-2-amino-2-carboxyethyl]-2-hydroxyphenyl]-4-hydroxyphenyl]propanoic acid;dihydrochloride |
| Molecular Weight (g/mol) | 433.28 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?