missing translation for 'onlineSavingsMsg'
Learn More

L-Kynurenine

Product Code. 15463009
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463009 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15463009 Supplier Thermo Scientific Alfa Aesar Supplier No. J60199.MF
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2922-83-0 | C10H12N2O3 | 208.22 g/mol

Intermediate in the breakdown of tryptophan

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2922-83-0
Color Yellow
Molecular Formula C10H12N2O3
MDL Number MFCD00069912
Quantity 50 mg
Synonym l-kynurenine, s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, kynurenine, l, 3-anthraniloyl-l-alanine, unii-02jw4j5r44, ccris 4425, 2s-2-amino-4-2-aminophenyl-4-oxobutanoic acid, quinurenine, kynurenin, s-alpha,2-diamino-gamma-oxobenzenebutanoic acid
InChI Key YGPSJZOEDVAXAB-QMMMGPOBSA-N
SMILES N[C@@H](CC(=O)C1=CC=CC=C1N)C(O)=O
IUPAC Name (2S)-2-amino-4-(2-aminophenyl)-4-oxobutanoic acid
Molecular Weight (g/mol) 208.22
PubChem CID 161166
ChEBI CHEBI:16946
Formula Weight 208.21
Physical Form Crystalline
Chemical Name or Material L-Kynurenine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.