missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Kynurenine, MedChemExpress
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
L-Kynurenine is a metabolite of the amino acid L-tryptophan. L-Kynurenine is an aryl hydrocarbon receptor agonist.
Specifications
Specifications
| Chemical Name or Material | L-Kynurenine |
| CAS | 2922-83-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C10H12N2O3 |
| Solubility Information | H2O : 20 mg/mL (96.06 mM; Need ultrasonic) ∣DMSO : 12.5 mg/mL (60.04 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | O=C(O)[C@@H](N)CC(C1=CC=CC=C1N)=O |
| Molecular Weight (g/mol) | 208.21 |
| Quantity | 50 mg |
| Formula Weight | 208.21 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction