missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Isovaline monohydrate, 99+%, (>99% ee)

Product Code. 10328663 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10328663 1 g 1g
10369440 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10328663 Supplier Thermo Scientific Chemicals Supplier No. 290050010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 595-40-4 | C5H11NO2 | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-40-4
Molecular Formula C5H11NO2
Molecular Weight (g/mol) 117.15
MDL Number MFCD00145255,MFCD00800497,MFCD00800497,MFCD00272968
InChI Key GCHPUFAZSONQIV-UHFFFAOYNA-N
Synonym d-isovaline, l-isovaline, s-2-amino-2-methylbutanoic acid, l-iso-valine, unii-ebv7h5w26h, ebv7h5w26h, isovaline, l, d--isovaline, 2s-2-amino-2-methylbutanoic acid, s-2-amino-2-methylbutyric acid
PubChem CID 6971276
IUPAC Name (2S)-2-amino-2-methylbutanoic acid
SMILES CCC(C)(N)C(O)=O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 305°C to 308°C
Color White
Enantiomeric Excess (GC):%gt; 99%
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass Bottle
Linear Formula CH3CH2CNH2(CH3)COOH·H2O
Quantity 1 g
Merck Index 15, 5280
Specific Rotation Condition +9.4° (25°C c=5,water)
Specific Rotation + 9.4
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol,slightly soluble in ether
Formula Weight 135.16
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(+)-Isovaline monohydrate, 99+% (>99% e.e.)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.