missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Isoleucyl-L-methionine TFA Salt, TRC
Specifications
Specifications
| Chemical Name or Material | L-Isoleucyl-L-methionine TFA Salt |
| Analyte or Component Names | L-Isoleucyl-L-methionine TFA Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H22 N2 O3 S . C2 H F3 O2 |
| Synonym | 1176: PN: EP2071334 SEQID: 1248 Claimed Protein, 1176: PN: WO2009077864 SEQID: 1248 Claimed Protein, 41: PN: EP2161028 PAGE: 9 Claimed Protein, 82: PN: WO2020097235 SEQID: 312 Claimed Protein |
| InChI Formula | InChI=1S/C11H22N2O3S.C2HF3O2/c1-4-7(2)9(12)10(14)13-8(11(15)16)5-6-17-3;3-2(4,5)1(6)7/h7-9H,4-6,12H2,1-3H3,(H,13,14)(H,15,16);(H,6,7)/t7-,8-,9-;/m0./s1 |
| Solution Type | Neat |
| SMILES | CC[C@H](C)[C@H](N)C(=O)N[C@@H](CCSC)C(=O)O.OC(=O)C(F)(F)F |
| IUPAC Name | (2S)-2-[[(2S,3S)-2-amino-3-methylpentanoyl]amino]-4-methylsulfanylbutanoic acid;2,2,2-trifluoroacetic acid |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?