missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucyl-L-methionine TFA Salt, TRC

Product Code. 30406834
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406834 500 mg 500mg
30393803 100 mg 100mg
30395402 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406834 Supplier Toronto Research Chemicals Supplier No. TRCI942083500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Isoleucyl-L-methionine TFA Salt
Analyte or Component Names L-Isoleucyl-L-methionine TFA Salt
Shipping Condition Room Temperature
Molecular Formula C11 H22 N2 O3 S . C2 H F3 O2
Synonym 1176: PN: EP2071334 SEQID: 1248 Claimed Protein, 1176: PN: WO2009077864 SEQID: 1248 Claimed Protein, 41: PN: EP2161028 PAGE: 9 Claimed Protein, 82: PN: WO2020097235 SEQID: 312 Claimed Protein
InChI Formula InChI=1S/C11H22N2O3S.C2HF3O2/c1-4-7(2)9(12)10(14)13-8(11(15)16)5-6-17-3;3-2(4,5)1(6)7/h7-9H,4-6,12H2,1-3H3,(H,13,14)(H,15,16);(H,6,7)/t7-,8-,9-;/m0./s1
Solution Type Neat
SMILES CC[C@H](C)[C@H](N)C(=O)N[C@@H](CCSC)C(=O)O.OC(=O)C(F)(F)F
IUPAC Name (2S)-2-[[(2S,3S)-2-amino-3-methylpentanoyl]amino]-4-methylsulfanylbutanoic acid;2,2,2-trifluoroacetic acid
Industry Type Pharmaceutical
Molecular Weight (g/mol) 376.392
Quantity 500 mg
Formula Weight 376.128
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.