missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine ethyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 15491919
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56782-52-6
Molecular Formula C8H18ClNO2
Molecular Weight (g/mol) 195.69
MDL Number MFCD09953729
InChI Key QQGRWNMNWONMOO-LEUCUCNGSA-N
Synonym l-isoleucine ethyl ester hydrochloride, h-ile-oet.hcl, ethyl l-isoleucinate hydrochloride, l-isoleucine ethyl ester hcl, 2s,3s-ethyl 2-amino-3-methylpentanoate hydrochloride, isoleucine, ethyl ester, hydrochloride, ethyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, l-isoleucine ethylester hcl, ksc497o6p, l-isoleucineethylesterhydrochloride
PubChem CID 6453390
SMILES Cl.CCOC(=O)[C@@H](N)[C@@H](C)CC

Specifications

Color White
Beilstein 4710859
IUPAC Name hydrogen ethyl (2S,3S)-2-amino-3-methylpentanoate chloride
Formula Weight 195.69
Percent Purity 98%
Quantity 1 g
Physical Form Powder
Chemical Name or Material L-Isoleucine ethyl ester hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'