missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Isoleucine-d10, TRC
Specifications
Specifications
| Chemical Name or Material | L-Isoleucine-d10 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6H3D10NO2 |
| Quantity | 100 mg |
| Synonym | (2S,3S)-2-Amino-3-methylpentanoic Acid-d10, (2S,3S)-α-Amino-β-methyl-n-valeric Acid-d10, (2S,3S)-α-Amino-β-methylvaleric Acid-d10, (S)-Isoleucine-d10, (S,S)-Isoleucine-d10, 1579-d10: PN: EP2071334 SEQID: 1669 Claimed Protein-d10, 1579: PN: WO2009077864 SEQID: 1669 Claimed Protein-d10, 2-Amino-3-methylvaleric Acid-d10, 2S,3S-Isoleucine-d10, 9: PN: US20090069547 PAGE: 10 Claimed Protein, Isoleucine-d10, L-(+)-Isoleucine, L-Ile-d10, 3-Methyl-erythro-L-norvaline-d10, NSC 46708-d10, (2S,3S)-2-Amino-3-methylpentanoic Acid-d10, [S-(R*,R*)]-2-Amino-3-methylpentanoic Acid-d10, erythro-L-Isoleucine-d10 |
| InChI Formula | InChI=1S/C21H23NO4/c1-3-13(2)19(20(23)24)22-21(25)26-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,13,18-19H,3,12H2,1-2H3,(H,22,25)(H,23,24)/t13-,19+/m0/s1/i1D3,2D3,3D2,13D,19D |
| SMILES | N[C@@](C(O)=O)([2H])[C@@]([2H])(C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])[2H] |
| Molecular Weight (g/mol) | 141.23 |
| Percent Purity | >95 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?