missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine, >99%, MP Biomedicals™

Product Code. 11378939 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378939 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11378939 Supplier MP Biomedicals Supplier No. 0210209291
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Specifications

CAS 73-32-5
Color White
Assay Percent Range >99%
Molecular Formula C6H13NO2
MDL Number MFCD00064222 MFCD00004268
Synonym l-isoleucine, isoleucine, 2s,3s-2-amino-3-methylpentanoic acid, s-isoleucine, s,s-isoleucine, 2s,3s-isoleucine, 2-amino-3-methylvaleric acid, erythro-l-isoleucine, l-+-isoleucine, l-ile
InChI Key AGPKZVBTJJNPAG-WHFBIAKZSA-N
SMILES CC[C@H](C)[C@H](N)C(O)=O
IUPAC Name (2S,3S)-2-amino-3-methylpentanoic acid
Molecular Weight (g/mol) 131.18
Quantity 1 kg
PubChem CID 6306
ChEBI CHEBI:17191
Formula Weight 131.2
Percent Purity 1.003
Physical Form Powder
Chemical Name or Material L-Isoleucine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.