missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ L-Isoleucine 7-amido-4-methylcoumarin trifluoroacetate salt

Product Code. 15490337
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 15490337

Brand: Alfa Aesar™ J64087.MF

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 191723-68-9
Color White
Molecular Formula C18H21F3N2O5
MDL Number MFCD00152116
Synonym l-isoleucine-7-amido-4-methylcoumarin trifluoroacetate salt, h-ile-amc tfa, l-isoleucine 7-amido-4-methylcoumarin trifluoroacetate salt, l-isolecucine 7-amido-4-methylcoumarin trifluoroacetate salt, l-isoleucine 7-amido-4-methylcoumarin, trifluoroacetate salt, trifluoroacetic acid-n-4-methyl-2-oxo-2h-1-benzopyran-7-yl-l-isoleucinamide 1/1, 2s,3s-2-amino-3-methyl-n-4-methyl-2-oxochromen-7-yl pentanamide; trifluoroacetic acid
InChI Key XPPJTDCCTDQORL-UXZWUECBSA-N
SMILES CCC(C)C(C(=O)NC1=CC2=C(C=C1)C(=CC(=O)O2)C)N.C(=O)(C(F)(F)F)O
IUPAC Name (2S,3S)-2-amino-3-methyl-N-(4-methyl-2-oxochromen-7-yl)pentanamide;2,2,2-trifluoroacetic acid
Molecular Weight (g/mol) 402.37
Quantity 50 mg
PubChem CID 24802239
Formula Weight 402.36
Physical Form Powder
Chemical Name or Material L-Isoleucine 7-amido-4-methylcoumarin trifluoroacetate salt
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.