missing translation for 'onlineSavingsMsg'
Learn More

L-Hydroxyproline,BioReagent, suitable for cell culture, MedChemExpress

Product Code. 16460508
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16460508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16460508 Supplier Medchem Express Supplier No. HY40135
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Hydroxyproline, one of the hydroxyproline (Hyp) isomers, is a useful chiral building block in the production of many pharmaceuticals.

L-Hydroxyproline, one of the hydroxyproline (Hyp) isomers, is a useful chiral building block in the production of many pharmaceuticals.

Specifications

Chemical Name or Material L-Hydroxyproline,BioReagent, suitable for cell culture
CAS 51-35-4
Color White
Purity Grade Notes Research
Molecular Formula C5H9NO3
Quantity 5 g
Solubility Information H2O : 50 mg/mL (381.30 mM; Need ultrasonic)
SMILES O=C(O)[C@H](C1)NC[C@@H]1O
Molecular Weight (g/mol) 131.13
Formula Weight 131.13
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.