missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine, TRC

Product Code. 30404408
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30404408 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30404408 Supplier Toronto Research Chemicals Supplier No. TRCH615010250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Homoserine
CAS 672-15-1
Purity Grade Notes HPLC
Molecular Formula C4 H9 N O3
Quantity 250 mg
Synonym 2(3H)-Furanone, 3-aminodihydro-, hydrobromide (1:1) (ACI), 2(3H)-Furanone, 3-aminodihydro-, hydrobromide (9CI), 2(3H)-Furanone, 3-aminodihydro-, hydrobromide, (±)- (ZCI), (±)-3-Aminodihydro-2(3H)-furanone hydrobromide, 2-Amino-4-butyrolactone hydrobromide, 2-Aminobutyrolactone hydrobromide, 3-Aminodihydro-2(3H)-furanone hydrobromide, 3-Aminooxolan-2-one hydrobromide, DL-Homoserine lactone hydrobromide, Alpha-amino-gamma-butyrolactone hydrobromide, Homoserine lactone hydrobromide, NSC 41120, α-Amino-γ-butyrolactone hydrobromide
InChI Formula InChI=1S/C4H9NO3/c5-3(1-2-6)4(7)8/h3,6H,1-2,5H2,(H,7,8)/t3-/m0/s1
SMILES N[C@@H](CCO)C(=O)O
IUPAC Name (2S)-2-amino-4-hydroxybutanoic acid
Molecular Weight (g/mol) 119.12
Formula Weight 119.0582
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.