missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine Lactone, Hydrochloride, TRC

Product Code. 30402942
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402942 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402942 Supplier Toronto Research Chemicals Supplier No. TRCH61510010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Homoserine Lactone, Hydrochloride
CAS 2185-03-7
Molecular Formula C4 H7 N O2 . Cl H
Quantity 10 g
Synonym 2(3H)-Furanone, 3-aminodihydro-, hydrochloride (1:1), (3S)-, 2(3H)-Furanone, 3-aminodihydro-, hydrochloride, (3S)- (9CI), 2(3H)-Furanone, 3-aminodihydro-, hydrochloride, (S)-, (3S)-3-Aminodihydro-2(3H)-furanone hydrochloride, (S)-Homoserine lactone hydrochloride, L-Homoserine lactone hydrochloride
InChI Formula InChI=1S/C4H7NO2.ClH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H/t3-;/m0./s1
SMILES Cl.N[C@H]1CCOC1=O
IUPAC Name (3S)-3-aminooxolan-2-one;hydrochloride
Molecular Weight (g/mol) 137.56
Formula Weight 137.0244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.