missing translation for 'onlineSavingsMsg'
Learn More

L-Homoserine Lactone Hydrobromide, TRC

Product Code. 30394346
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394346 1 g 1g
30407655 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394346 Supplier Toronto Research Chemicals Supplier No. TRCH6151051G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Homoserine Lactone Hydrobromide
CAS 15295-77-9
Molecular Formula C4 H7 N O2 . HBr
Quantity 1 g
Synonym (3S)-3-Aminodihydro-2(3H)-furanone Hydrobromide, (S)-3-Aminodihydro-2(3H)-furanone Hydrobromide, L-3-Aminodihydro-2(3H)-furanone Hydrobromide, (S)-(-)-α-Aminobutyrolactone Hydrobromide, (S)-2-Amino-4-butyrolactone Hydrobromide, (S)-3-Aminodihydrofuran-2(3H)-one Hydrobromide
InChI Formula InChI=1S/C4H7NO2.BrH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H/t3-;/m0./s1
SMILES N[C@@H]1C(=O)OCC1.Br
IUPAC Name (3S)-3-aminotetrahydrofuran-2-one;hydrobromide
Molecular Weight (g/mol) 182.01582
Formula Weight 180.9738
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.