Learn More
L-Homoserine, 99%
Descrizione
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Homoserine is used in the biosynthesis of methionine, threonine and isoleucine.
Solubility
Soluble in water (1100 mg/ml at 30°C), and methanol (Sparingly). Insoluble in ethanol.
Notes
Store in a cool, dry conditions in well sealed container. Incompatible with oxidizing agents. Store at room temperature.
Specifica
Specifications
| Melting Point | ∽203°C (decomposition) |
| Beilstein | 1721681 |
| Merck Index | 14,4739 |
| Solubility Information | Soluble in water (1100mg/ml at 30°C),and methanol (Sparingly). Insoluble in ethanol. |
| Quantity | 250 mg |
| Formula Weight | 119.12 |
| Optical Rotation | −9° (c=2 in Water) |
| Percent Purity | 99% |
| Chemical Name or Material | L-Homoserine |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.