missing translation for 'onlineSavingsMsg'
Learn More

L-beta-Homoproline hydrochloride, 98%

Product Code. 10745915 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10745915 1 g Glass Bottle 1g
10535144 250 mg Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10745915 Supplier Thermo Scientific Acros Supplier No. 426170010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 53912-85-9 | C6H12ClNO2 | 165.617 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53912-85-9
Molecular Formula C6H12ClNO2
Molecular Weight (g/mol) 165.617
MDL Number MFCD07363484
InChI Key VQDACVOAOJQTPR-JEDNCBNOSA-N
Synonym s-2-pyrrolidin-2-yl acetic acid hydrochloride, l-beta-homoproline hydrochloride, l-beta-homoproline hcl, 2-2s-pyrrolidin-2-yl acetic acid hydrochloride, 2-pyrrolidineacetic acid, hydrochloride, 2s, s-2-2-pyrrolidinyl acetic acid hydrochloride, s-3-pyrrolidinecarboxylic acid hydrochloride, 2s-pyrrolidin-2-ylacetic acid hydrochloride, l-beta-homoproline-hcl, s-3-pyrrol-idinecarboxylic acid hydrochloride
PubChem CID 2761540
IUPAC Name 2-[(2S)-pyrrolidin-2-yl]acetic acid;hydrochloride
SMILES C1CC(NC1)CC(=O)O.Cl

Specifications

Color White
Infrared Spectrum Authentic
NMR Consistent with structure
Assay Percent Range 98%
Packaging Glass Bottle
Specific Rotation +31° to +35° (20°C, 589nm) (c=1, H2O)
Quantity 1 g
Formula Weight 165.62
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material L-β-Homoproline hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L-beta-Homoproline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.