missing translation for 'onlineSavingsMsg'
Learn More

L-Homocystine-3,3,3’,3’,4,4,4’,4’-d8, TRC

Product Code. 30397378
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397378 1 mg -
30399831 2.5 mg 2.5mg
30404103 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30397378 Supplier Toronto Research Chemicals Supplier No. TRCH5913621MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 182755-41-5
Molecular Formula C8 D8 H8 N2 O4 S2
Molecular Weight (g/mol) 276.4
Synonym (2S,2'S)-4,4'-Dithiobis[2-aminobutanoic-3,3,4,4-d4 Acid], [S-(R*,R*)]-4,4'-Dithiobis[2-amino-butanoic-3,3,4,4-d4 Acid
IUPAC Name (2S)-2-amino-4-[[(3S)-3-amino-3-carboxy-1,1,2,2-tetradeuteriopropyl]disulfanyl]-3,3,4,4-tetradeuteriobutanoic acid
SMILES [2H]C([2H])(SSC([2H])([2H])C([2H])([2H])[C@H](N)C(=O)O)C([2H])([2H])[C@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C8H16N2O4S2/c9-5(7(11)12)1-3-15-16-4-2-6(10)8(13)14/h5-6H,1-4,9-10H2,(H,11,12)(H,13,14)/t5-,6-/m0/s1/i1D2,2D2,3D2,4D2
Quantity 1 mg
Formula Weight 276.1054
Shipping Condition Room Temperature
Chemical Name or Material L-Homocystine-3,3,3',3',4,4,4',4'-d8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.