missing translation for 'onlineSavingsMsg'
Learn More

L-Homocysteinesulfinic Acid, TRC

Product Code. 30408361
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30408361 5 mg 5mg
30400450 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30408361 Supplier Toronto Research Chemicals Supplier No. TRCH6160005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2686-70-6
Molecular Formula C4H9NO4S
Molecular Weight (g/mol) 167.18
Synonym 2-Amino-4-sulfinobutanoic Acid, L-2-Amino-4-sulfinobutyric Acid, (2S)-2-Amino-4-sulfinobutanoic Acid, L-Homocysteine Sulfinate
IUPAC Name (2S)-2-amino-4-sulfinobutanoic acid
SMILES N[C@@H](CCS(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C4H9NO4S/c5-3(4(6)7)1-2-10(8)9/h3H,1-2,5H2,(H,6,7)(H,8,9)/t3-/m0/s1
Quantity 5 mg
Formula Weight 167.0252
Shipping Condition Room Temperature
Chemical Name or Material L-Homocysteinesulfinic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.