missing translation for 'onlineSavingsMsg'
Learn More

L-Homoarginine-d4 Dihydrochloride, TRC

Product Code. 30402715
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402715 10 mg 10mg
30403516 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30402715 Supplier Toronto Research Chemicals Supplier No. TRCH58500210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C7H14D4Cl2N4O2
Molecular Weight (g/mol) 192.2623646
Synonym N6-(Aminoiminomethyl)-L-lysine-d4 Dihydrochloride, 2-Amino-6-guanidinohexanoic Acid-d4 Dihydrochloride, NSC 145416-d4
SMILES NC(NCC([2H])(C([2H])(C[C@@H](C(O)=O)N)[2H])[2H])=N.[2HCl]

Specifications

InChI Formula InChI=1S/C7H16N4O2.2ClH/c8-5(6(12)13)3-1-2-4-11-7(9)10;;/h5H,1-4,8H2,(H,12,13)(H4,9,10,11);2*1H/t5-;;/m0../s1/i3D,5D;;/hD2/t3?,5-;;
Quantity 10 mg
Formula Weight 264.11
Shipping Condition Room Temperature
Chemical Name or Material L-Homoarginine-d4 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.