missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine (White Crystals or Crystalline Powder), Fisher BioReagents

Product Code. 10254823
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10254823 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10254823 Supplier Fisher BioReagents Supplier No. BP382100

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
This amino acid is suitable for use in tissue culture systems requiring additives.
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 282°C
Ammonium 0.02% max.
CAS 71-00-1
Arsenic (As) 1ppm max.
pH 7.0 to 8.5
Color White
Heavy Metals (as Pb) 10ppm max.
Ignition Residue 0.10% max.
Identification Pass Test
Assay Percent Range 98.5 to 101.0 %
Loss on Drying 0.20% max.
Molecular Formula C6H9N3O2
Packaging Glass Bottle
MDL Number MFCD00064315
Specific Rotation 12.0 to 13.0° (+ or -)
Merck Index 15, 4758
State of Solution (%T) 98.0% min.
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
Alpha Vector Histidine
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
SMILES NC(CC1=CN=CN1)C(O)=O
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid
Molecular Weight (g/mol) 155.16
Quantity 100 g
PubChem CID 6274
ChEBI CHEBI:15971
Formula Weight 155.16
Percent Purity 98.5 to 101.0%
Physical Form Solid
Chemical Name or Material L-Histidine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.