missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine Monohydrochloride Monohydrate, TRC

Product Code. 30136554
Change view
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136554 10 g 10g
30006312 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30136554 Supplier Toronto Research Chemicals Supplier No. TRCH45603810G
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6 H9 N3 O2 . Cl H . H2 O
Molecular Weight (g/mol) 209.63
Synonym Histidine hydrochloride monohydrate, (S)-2-Amino-3-(imidazol-4-yl)propanoic acid hydrochloride monohydrate, L-Histidine, hydrochloride, hydrate (1:1:1) (ACI), L-Histidine, monohydrochloride, monohydrate (9CI), Histidine, monohydrochloride, monohydrate, L- (8CI), L-Histidine hydrochloride monohydrate, L-Histidine monohydrate monohydrochloride, (2S)-2-Amino-3-(1H-imidazol-4-yl)propanoic acid monohydrate monohydrochloride
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid;hydrate;hydrochloride
SMILES O.Cl.N[C@@H](Cc1c[nH]cn1)C(=O)O

Specifications

InChI Formula InChI=1S/C6H9N3O2.ClH.H2O/c7-5(6(10)11)1-4-2-8-3-9-4;;/h2-3,5H,1,7H2,(H,8,9)(H,10,11);1H;1H2/t5-;;/m0../s1
Quantity 10 g
Formula Weight 209.0567
Shipping Condition Room Temperature
Chemical Name or Material Histidine hydrochloride monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.