missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine Monohydrochloride Monohydrate, ≥98%, Thermo Scientific™

Product Code. 15885158 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15885158 100 g Plastic Bottle 100g
15805308 1 kg Plastic Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15885158 Supplier Thermo Scientific Supplier No. J17080.22

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1 h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.H2O, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Melting Point ∼240°C (decomposition)
Packaging Plastic Bottle
Linear Formula (C3H3N2)CH2CH(NH2)COOH.HCl.H2O
Beilstein 4168261
Quantity 100 g
Formula Weight 209.63
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine Monohydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.