missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine methyl ester dihydrochloride, 98+%

Product Code. 11429543
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11429543 25 g 25g
11419543 5 g 5g
11439543 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11429543 Supplier Thermo Scientific Alfa Aesar Supplier No. A12386.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7389-87-9 | C7H11ClN3O2 | 204.63 g/mol

L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7389-87-9
Molecular Formula C7H11ClN3O2
Molecular Weight (g/mol) 204.63
MDL Number MFCD00012701
InChI Key VXXIPPPPKNWFLK-UHFFFAOYNA-N
Synonym l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
PubChem CID 2723645
SMILES [Cl].COC(=O)C(N)CC1=CN=CN1

Specifications

Melting Point 198°C (decomposition)
Beilstein 3572010
Solubility Information Soluble in dimethyl sulfoxide,methanol and water.
IUPAC Name methyl (2S)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
Quantity 25 g
Formula Weight 242.11
Optical Rotation +10° (c=2 in water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Histidine methyl ester dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.