missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Histidine methyl ester dihydrochloride, 97%

Product Code. 10265060 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265060 25 g Glass bottle 25g
10118043 5 g Glass bottle 5g
10520391 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10265060 Supplier Thermo Scientific Chemicals Supplier No. 120820250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7389-87-9 | C7H11ClN3O2 | 204.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7389-87-9
Molecular Formula C7H11ClN3O2
Molecular Weight (g/mol) 204.63
MDL Number MFCD00012701
InChI Key VXXIPPPPKNWFLK-UHFFFAOYNA-N
Synonym l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
PubChem CID 2723645
IUPAC Name methyl (2S)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
SMILES [Cl].COC(=O)C(N)CC1=CN=CN1

Specifications

Melting Point 207.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 25 g
Beilstein 25, 515
Specific Rotation Condition +15° (24°C c=1,MeOH)
Specific Rotation + 15.00
Formula Weight 242.11
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(+)-Histidine methyl ester dihydrochloride, 97%
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Histidine methyl ester dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.