missing translation for 'onlineSavingsMsg'
Learn More

Melford L-Histidine HCl

Product Code. 30018347
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018347 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30018347 Supplier Melford Supplier No. H750801000.0
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

L-Histidine Hydrochloride Monohydrate is a compound commonly used in life science research, particularly in biochemistry and molecular biology.

L-Histidine Hydrochloride Monohydrate is a compound commonly used in life science research, particularly in biochemistry and molecular biology. It is the hydrochloride salt form of L-histidine, an essential amino acid that plays crucial roles in protein synthesis, enzyme catalysis, and neurotransmitter regulation. The monohydrate aspect indicates that it contains one molecule of water per molecule of the compound.

In research, L-Histidine Hydrochloride Monohydrate is often utilized in cell culture media to support cell growth and viability, as well as in protein purification and crystallization processes.

Its availability in a stable, crystalline form makes it valuable for experimental reproducibility and accuracy in various biological assays and investigations.

Specifications

Chemical Name or Material L-Histidine HCl
CAS 5934-29-2
Molecular Formula C6H9N3O2-HCl-H20
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.