missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, free base, ≥99%, MP Biomedicals™

Product Code. p-4404399 Shop All MP Biomedicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
This item is not returnable. View return policy

Product Code. 11384419

Brand: MP Biomedicals 0210195425

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Precursor of histamine by action of histidine decarboxylase

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Quantity 25 g
Formula Weight 155.16
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.