missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, free base, ≥99%, MP Biomedicals™

Product Code. 11314429 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
Unit Size:
100g
1kg
25g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11314429 1 kg 1kg
11384419 25 g 25g
11394419 100 g 100g
11304429 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11314429 Supplier MP Biomedicals Supplier No. 0210195491
  • ONLINE EXCLUSIVE PRICE
    £1135.00
    £1061.55 / 1kg
    Save £73.45
    6% Off
View all promotions
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Precursor of histamine by action of histidine decarboxylase

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Color White
Assay Percent Range ≥99%
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
Quantity 1 kg
Formula Weight 155.16
Physical Form Crystalline Powder
Chemical Name or Material L-Histidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.